Gene Transcripts
- Gene Symbol
- Gm14634
- Entrez Gene ID
- 552913
- Gene Name
- predicted gene 14634
- Gene Aliases
- OTTMUSG00000017535
- Chromosome Location
- Chr. X: 12762278 - 12821492 on Build GRCm38
- UniGene ID
- Mm.216846
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045852.1 |
|
3 |
872 |
|
GenBank mRNA |
AK036020.1 |
- |
3 |
873 |
|
AK132710.1 |
- |
3 |
607 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CJ046751.1 |
- |
3 |
607 |
|
Chr. X - 12762544 - 12817291 [+] |
View Full Product Details