Gene Transcripts

Gene Symbol
Klhl22
Entrez Gene ID
224023
Gene Name
kelch-like 22
Gene Aliases
2610318I18Rik, Kelchl
Chromosome Location
Chr. 16: 17759090 - 17793382 on Build GRCm38
UniGene ID
Mm.282807
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK012046.1 - 1 1971


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CD350527.1 - 1 571 Chr. 16 - 17761070 - 17761794 [+]

View Full Product Details