Gene Transcripts
- Gene Symbol
- Rgs22
- Entrez Gene ID
- 626596
- Gene Name
- regulator of G-protein signalling 22
- Gene Aliases
- Chromosome Location
- Chr. 15: 35960511 - 36140630 on Build GRCm38
- UniGene ID
- Mm.321798
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK158239.1 |
- |
2 |
264 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY753024.1 |
- |
2 |
264 |
|
Chr. 15 - 36139231 - 36140385 [-] |
View Full Product Details