Gene Transcripts
- Gene Symbol
- B230398E01Rik
- Entrez Gene ID
- 109334
- Gene Name
- RIKEN cDNA B230398E01 gene
- Gene Aliases
- 6720476P12Rik
- Chromosome Location
- Chr. 3: 94959674 - 94986687 on Build GRCm38
- UniGene ID
- Mm.470046
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK046489.1 |
- |
|
1192 |
|
View Full Product Details