Gene Transcripts

Gene Symbol
4930571O06Rik
Entrez Gene ID
78048
Gene Name
RIKEN cDNA 4930571O06 gene
Gene Aliases
Chromosome Location
Chr. 8: 113517282 - 113550808 on Build GRCm38
UniGene ID
Mm.252797
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110476.1 2 133
GenBank mRNA AK019805.1 - 2 134


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY715894.1 - 2 134 Chr. 8 - 113517282 - 113550810 [+]

View Full Product Details