Gene Transcripts
- Gene Symbol
- Gm31938
- Entrez Gene ID
- 102634330
- Gene Name
- predicted gene, 31938
- Gene Aliases
- Chromosome Location
- Chr. 12: 4523853 - 4529122 on Build GRCm38
- UniGene ID
- Mm.402324
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK077964.1 |
- |
3 |
482 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB621333.1 |
- |
3 |
478 |
|
Chr. 12 - 4524115 - 4527964 [+] |
View Full Product Details