Gene Transcripts
- Gene Symbol
- 4930461G14Rik
- Entrez Gene ID
- 74879
- Gene Name
- RIKEN cDNA 4930461G14 gene
- Gene Aliases
- Chromosome Location
- Chr. 9: 58455173 - 58469623 on Build GRCm38
- UniGene ID
- Mm.416924
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040736.1 |
|
3 |
548 |
|
GenBank mRNA |
AK015493.1 |
- |
3 |
548 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AI046901.1 |
- |
2 |
324 |
|
Chr. 9 - 58466841 - 58469401 [+] |
GenBankmRNA |
BY715041.1 |
- |
3 |
548 |
|
Chr. 9 - 58455173 - 58469618 [+] |
View Full Product Details