Gene Transcripts
- Gene Symbol
- 5430410E06Rik
- Entrez Gene ID
- 71413
- Gene Name
- RIKEN cDNA 5430410E06 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.439675
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017289.1 |
- |
1 |
918 |
|
View Full Product Details