Gene Transcripts
- Gene Symbol
- Plxdc2
- Entrez Gene ID
- 67448
- Gene Name
- plexin domain containing 2
- Gene Aliases
- 1200007L24Rik, 5430431D22Rik, AI646728, AU022916, AV231634, Tem7r
- Chromosome Location
- Chr. 2: 16356210 - 16755839 on Build GRCm38
- UniGene ID
- Mm.313938
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017369.1 |
- |
1 |
267 |
|
View Full Product Details