Gene Transcripts
- Gene Symbol
- Tenm2
- Entrez Gene ID
- 23964
- Gene Name
- teneurin transmembrane protein 2
- Gene Aliases
- Odz2, Odz3, Ten-2, Ten-m2
- Chromosome Location
- Chr. 11: 36006656 - 37236808 on Build GRCm38
- UniGene ID
- Mm.439889
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020393.1 |
- |
1 |
380 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY721501.1 |
- |
1 |
380 |
|
Chr. 11 - 36671960 - 36672602 [-] |
View Full Product Details