Gene Transcripts
- Gene Symbol
- Gm31121
- Entrez Gene ID
- 102633249
- Gene Name
- predicted gene, 31121
- Gene Aliases
- Chromosome Location
- Chr. 3: 149562882 - 149660394 on Build GRCm38
- UniGene ID
- Mm.428127
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK139864.1 |
- |
5 |
482 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY725998.1 |
- |
5 |
479 |
|
Chr. 3 - 149563639 - 149660393 [-] |
View Full Product Details