Gene Transcripts
- Gene Symbol
- 1700027H10Rik
- Entrez Gene ID
- 72274
- Gene Name
- RIKEN cDNA 1700027H10 gene
- Gene Aliases
- Chromosome Location
- Chr. 3: 45416584 - 45439309 on Build GRCm38
- UniGene ID
- Mm.396202
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040594.1 |
|
1 |
40 |
|
GenBank mRNA |
AK006417.1 |
- |
1 |
41 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY706472.1 |
- |
1 |
38 |
|
Chr. 3 - 45416584 - 45439307 [-] |
View Full Product Details