Gene Transcripts
- Gene Symbol
- Gm30332
- Entrez Gene ID
- 102632192
- Gene Name
- predicted gene, 30332
- Gene Aliases
- Chromosome Location
- Chr. 6: 138443973 - 138461615 on Build GRCm38
- UniGene ID
- Mm.417247
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK146614.1 |
- |
5 |
2131 |
|
AK169220.1 |
- |
5 |
2131 |
|
View Full Product Details