Gene Transcripts
- Gene Symbol
- Ppp2r5c
- Entrez Gene ID
- 26931
- Gene Name
- protein phosphatase 2, regulatory subunit B', gamma
- Gene Aliases
- 2610043M05Rik, 2700063L20Rik, AI060890, AW545884, C85228, D12Bwg0916e, mKIAA0044
- Chromosome Location
- Chr. 12: 110447179 - 110583075 on Build GRCm38
- UniGene ID
- Mm.240396
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK034227.1 |
- |
1 |
585 |
|
View Full Product Details