Gene Transcripts
- Gene Symbol
- Fem1c
- Entrez Gene ID
- 240263
- Gene Name
- fem-1 homolog c (C.elegans)
- Gene Aliases
- 2610312A07Rik, 3632443A22Rik, AI646567
- Chromosome Location
- Chr. 18: 46504606 - 46525971 on Build GRCm38
- UniGene ID
- Mm.38249
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK011987.1 |
- |
|
189 |
|
View Full Product Details