Gene Transcripts
- Gene Symbol
- 1700025M24Rik
- Entrez Gene ID
- 72276
- Gene Name
- RIKEN cDNA 1700025M24 gene
- Gene Aliases
- Chromosome Location
- Chr. 5: 73268580 - 73284184 on Build GRCm38
- UniGene ID
- Mm.271215
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040687.1 |
|
3 |
218 |
|
GenBank mRNA |
BC048655.1 |
- |
3 |
215 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CA465171.1 |
- |
|
218 |
|
Chr. - - [-] |
View Full Product Details