Gene Transcripts
- Gene Symbol
- Gm20187
- Entrez Gene ID
- 100504347
- Gene Name
- predicted gene, 20187
- Gene Aliases
- Chromosome Location
- Chr. 12: 27090237 - 27114995 on Build GRCm38
- UniGene ID
- Mm.130500
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045052.1 |
|
1 |
35 |
|
GenBank mRNA |
AK084401.1 |
- |
1 |
36 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB658287.1 |
- |
1 |
34 |
|
Chr. 12 - 27096756 - 27114994 [-] |
View Full Product Details