Gene Transcripts
- Gene Symbol
- Vangl2
- Entrez Gene ID
- 93840
- Gene Name
- vang-like 2 (van gogh, Drosophila)
- Gene Aliases
- C530001F03Rik, Lp, Lpp1, Ltap, Vang1l2, loop-tail, ska17, ska<m17Jus>, stbm, strabismus
- Chromosome Location
- Chr. 1: 172004360 - 172027314 on Build GRCm38
- UniGene ID
- Mm.36148
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK082938.1 |
- |
|
748 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX521967.1 |
- |
1 |
130 |
|
Chr. 1 - 172001257 - 172001771 [-] |
View Full Product Details