Gene Transcripts
- Gene Symbol
- Ilf3
- Entrez Gene ID
- 16201
- Gene Name
- interleukin enhancer binding factor 3
- Gene Aliases
- MBII-26, MPHOSPH4, NF90, NFAR
- Chromosome Location
- Chr. 9: 21367871 - 21405363 on Build GRCm38
- UniGene ID
- Mm.440026
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK052536.1 |
- |
1 |
2960 |
|
View Full Product Details