Gene Transcripts

Gene Symbol
Gnb1l
Entrez Gene ID
13972
Gene Name
guanine nucleotide binding protein (G protein), beta polypeptide 1-like
Gene Aliases
ESTM55, Gm16314, Me49f07, OTTMUSG00000033458, Wdr14, Wdvcf
Chromosome Location
Chr. 16: 18498021 - 18566680 on Build GRCm38
UniGene ID
Mm.236139
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC029862.1 - 3 530


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BE571862.1 - 3 510 Chr. 16 - 18498921 - 18501487 [+]

View Full Product Details