Gene Transcripts
- Gene Symbol
- 2210009P08Rik
- Entrez Gene ID
- 72292
- Gene Name
- RIKEN cDNA 2210009P08 gene
- Gene Aliases
- AV074459
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK008691.1 |
- |
1 |
198 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY708583.1 |
- |
1 |
198 |
|
Chr. 16 - 97466484 - 97466948 [-] |
View Full Product Details