Gene Transcripts
- Gene Symbol
- Gm15527
- Entrez Gene ID
- 100502661
- Gene Name
- predicted gene 15527
- Gene Aliases
- OTTMUSG00000023238
- Chromosome Location
- Chr. 6: 54241046 - 54263118 on Build GRCm38
- UniGene ID
- Mm.446956
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK080009.1 |
- |
|
62 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB636958.1 |
- |
1 |
62 |
|
Chr. 6 - 54248732 - 54279076 [-] |
View Full Product Details