Gene Transcripts
- Gene Symbol
- Gm16239
- Entrez Gene ID
- 102637685
- Gene Name
- predicted gene 16239
- Gene Aliases
- OTTMUSG00000032481
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.404895
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK139491.1 |
- |
3 |
489 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY725311.1 |
- |
3 |
485 |
|
Chr. 10 - 99181645 - 99184607 [-] |
View Full Product Details