Gene Transcripts
- Gene Symbol
- 4833422M21Rik
- Entrez Gene ID
- 75768
- Gene Name
- RIKEN cDNA 4833422M21 gene
- Gene Aliases
- 4933406G12Rik
- Chromosome Location
- Chr. 15: 96231870 - 96254684 on Build GRCm38
- UniGene ID
- Mm.212701
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016691.1 |
- |
3 |
668 |
|
AK084200.1 |
- |
3 |
697 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY716377.1 |
- |
3 |
668 |
|
Chr. 15 - 96232368 - 96241789 [-] |
View Full Product Details