Gene Transcripts
- Gene Symbol
- Gm17745
- Entrez Gene ID
- 432488
- Gene Name
- predicted gene, 17745
- Gene Aliases
- Chromosome Location
- Chr. 10: 93336232 - 93348879 on Build GRCm38
- UniGene ID
- Mm.32833
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038014.1 |
|
3 |
705 |
|
GenBank mRNA |
AK144329.1 |
- |
3 |
706 |
|
- Gene Symbol
- Gm3546
- Entrez Gene ID
- 100041854
- Gene Name
- predicted gene 3546
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.32833
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AI450451.1 |
- |
|
235 |
|
View Full Product Details