Gene Transcripts
- Gene Symbol
- 1700063O14Rik
- Entrez Gene ID
- 73452
- Gene Name
- RIKEN cDNA 1700063O14 gene
- Gene Aliases
- Chromosome Location
- Chr. 5: 91766242 - 91767300 on Build GRCm38
- UniGene ID
- Mm.396233
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045383.1 |
|
3 |
295 |
|
GenBank mRNA |
AK006875.1 |
- |
3 |
295 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY706861.1 |
- |
2 |
294 |
|
Chr. 5 - 91766242 - 91767128 [-] |
View Full Product Details