Gene Transcripts

Gene Symbol
Adamts3
Entrez Gene ID
330119
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 3
Gene Aliases
1100001H14Rik, 6330442E02Rik
Chromosome Location
Chr. 5: 89673841 - 89883669 on Build GRCm38
UniGene ID
Mm.133062
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK003167.1 - 1 154


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB193360.2 - 1 208 Chr. 5 - 89674864 - 89675422 [-]

View Full Product Details