Gene Transcripts
- Gene Symbol
- Mllt10
- Entrez Gene ID
- 17354
- Gene Name
- myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 10
- Gene Aliases
- Af10, B130021D15Rik, D630001B22Rik, mKIAA4140
- Chromosome Location
- Chr. 2: 18055175 - 18212390 on Build GRCm38
- UniGene ID
- Mm.209175
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK085252.1 |
- |
3 |
2906 |
|
AK085575.1 |
- |
1 |
1931 |
|
View Full Product Details