Gene Transcripts
- Gene Symbol
- 5830420C07Rik
- Entrez Gene ID
- 77599
- Gene Name
- RIKEN cDNA 5830420C07 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.438573
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020016.1 |
- |
1 |
2047 |
|
View Full Product Details