Gene Transcripts
- Gene Symbol
- 4933407E24Rik
- Entrez Gene ID
- 108809
- Gene Name
- RIKEN cDNA 4933407E24 gene
- Gene Aliases
- Chromosome Location
- Chr. 4: 124569179 - 124575538 on Build GRCm38
- UniGene ID
- Mm.250313
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045819.1 |
|
3 |
1809 |
|
GenBank mRNA |
AK160260.1 |
- |
3 |
1799 |
|
View Full Product Details