Gene Transcripts
- Gene Symbol
- Hottip
- Entrez Gene ID
- 791364
- Gene Name
- Hoxa distal transcript antisense RNA
- Gene Aliases
- ENSMUSG00000055408, Gm15053, OTTMUSG00000018874
- Chromosome Location
- Chr. 6: 52262775 - 52267603 on Build GRCm38
- UniGene ID
- Mm.97466
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110441.1 |
|
2 |
2756 |
|
NR_110442.1 |
|
2 |
2202 |
|
GenBank mRNA |
BC046496.1 |
- |
2 |
2202 |
|
View Full Product Details