Gene Transcripts
- Gene Symbol
- 4933400F21Rik
- Entrez Gene ID
- 74403
- Gene Name
- RIKEN cDNA 4933400F21 gene
- Gene Aliases
- 1700091C04Rik, 4930565K10Rik
- Chromosome Location
- Chr. 1: 89676507 - 89683892 on Build GRCm38
- UniGene ID
- Mm.444291
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016229.1 |
- |
|
657 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715838.1 |
- |
5 |
654 |
|
Chr. 1 - 89650641 - 89683908 [-] |
View Full Product Details