Gene Transcripts
- Gene Symbol
- D930030I03Rik
- Entrez Gene ID
- 320544
- Gene Name
- RIKEN cDNA D930030I03 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.20047
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK086460.1 |
- |
1 |
168 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB663080.1 |
- |
1 |
168 |
|
Chr. 16 - 38451982 - 38453249 [+] |
View Full Product Details