Gene Transcripts

Gene Symbol
Foxp1
Entrez Gene ID
108655
Gene Name
forkhead box P1
Gene Aliases
3110052D19Rik, 4932443N09Rik, AI461938, AW494214
Chromosome Location
Chr. 6: 98925342 - 99523024 on Build GRCm38
UniGene ID
Mm.234965
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK014203.1 - 1 167
AK033622.1 - 1 1678
AK157606.1 - 1 1711
AK169176.1 - 1 2487


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DT906875.1 - 145 Chr. - - [-]
GenBankmRNA BI158141.1 - 1 22 Chr. 6 - 98927082 - 98927644 [-]
GenBankmRNA BY760851.1 - 1 156 Chr. 6 - 98927102 - 98927779 [-]
GenBankmRNA DT911052.1 - 1 198 Chr. 6 - 98927381 - 98927820 [-]
GenBankmRNA CB202659.1 - 246 Chr. - - [-]
GenBankmRNA AV226704.2 - 1 174 Chr. 6 - 98927102 - 98927795 [-]
GenBankmRNA AA125102.1 - 1 185 Chr. 6 - 98927380 - 98927807 [-]
GenBankmRNA EH094178.1 - 1 151 Chr. 6 - 98927289 - 98927772 [-]
GenBankmRNA BF658606.1 - 1 322 Chr. 6 - 98927374 - 98927941 [-]
GenBankmRNA CJ232680.1 - 1 180 Chr. 6 - 98927097 - 98927803 [-]
GenBankmRNA BF658439.1 - 1 37 Chr. 6 - 98927174 - 98927659 [-]
GenBankmRNA BG095300.1 - 1 188 Chr. 6 - 98927449 - 98927810 [-]

View Full Product Details