Gene Transcripts
- Gene Symbol
- 1700089I07Rik
- Entrez Gene ID
- 78630
- Gene Name
- RIKEN cDNA 1700089I07 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.71125
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018915.1 |
- |
1 |
86 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY707091.1 |
- |
1 |
86 |
|
Chr. 2 - 154782332 - 154782683 [+] |
View Full Product Details