Gene Transcripts
- Gene Symbol
- 4930502M04Rik
- Entrez Gene ID
- 74974
- Gene Name
- RIKEN cDNA 4930502M04 gene
- Gene Aliases
- Chromosome Location
- Chr. 3: 130131505 - 130151534 on Build GRCm38
- UniGene ID
- Mm.91728
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015682.1 |
- |
|
683 |
|
View Full Product Details