Gene Transcripts
- Gene Symbol
- 4930550I04Rik
- Entrez Gene ID
- 75262
- Gene Name
- RIKEN cDNA 4930550I04 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.84630
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016087.1 |
- |
2 |
481 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715689.1 |
- |
2 |
481 |
|
Chr. 2 - 72629018 - 72630432 [-] |
View Full Product Details