Gene Transcripts
- Gene Symbol
- 4930550C17Rik
- Entrez Gene ID
- 75245
- Gene Name
- RIKEN cDNA 4930550C17 gene
- Gene Aliases
- Chromosome Location
- Chr. 13: 56238079 - 56243951 on Build GRCm38
- UniGene ID
- Mm.159475
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016084.1 |
- |
|
666 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715686.1 |
- |
5 |
670 |
|
Chr. 13 - 56222836 - 56243940 [-] |
View Full Product Details