Gene Transcripts
- Gene Symbol
- 2300004M11Rik
- Entrez Gene ID
- 70084
- Gene Name
- RIKEN cDNA 2300004M11 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK009029.1 |
- |
1 |
380 |
|
View Full Product Details