Gene Transcripts

Gene Symbol
Ttc7
Entrez Gene ID
225049
Gene Name
tetratricopeptide repeat domain 7
Gene Aliases
1110035E02Rik, 1700007L07Rik, Ttc7a, fsn, hea
Chromosome Location
Chr. 17: 87282082 - 87381770 on Build GRCm38
UniGene ID
Mm.77396
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK005733.1 - 2 549


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY705898.1 - 2 544 Chr. 17 - 87329789 - 87332049 [-]

View Full Product Details