Gene Transcripts
- Gene Symbol
- C330013F16Rik
- Entrez Gene ID
- 78526
- Gene Name
- RIKEN cDNA C330013F16 gene
- Gene Aliases
- Chromosome Location
- Chr. X: 139240226 - 139357738 on Build GRCm38
- UniGene ID
- Mm.439073
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045455.1 |
|
20 |
3146 |
|
GenBank mRNA |
AK133643.1 |
- |
20 |
3147 |
|
View Full Product Details