Gene Transcripts
- Gene Symbol
- 1700029M03Rik
- Entrez Gene ID
- 69476
- Gene Name
- RIKEN cDNA 1700029M03 gene
- Gene Aliases
- Chromosome Location
- Chr. 1: 164960664 - 164961400 on Build GRCm38
- UniGene ID
- Mm.67177
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076808.1 |
- |
|
195 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB615319.1 |
- |
2 |
191 |
|
Chr. 1 - 164960924 - 164962825 [+] |
GenBankmRNA |
BB013992.2 |
- |
2 |
195 |
|
Chr. 1 - 164960921 - 164962822 [+] |
View Full Product Details