Gene Transcripts
- Gene Symbol
- 1700093C20Rik
- Entrez Gene ID
- 76623
- Gene Name
- RIKEN cDNA 1700093C20 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.460423
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK044044.1 |
- |
1 |
338 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB273442.2 |
- |
1 |
381 |
|
Chr. 8 - 69749325 - 69749952 [-] |
GenBankmRNA |
BU603970.1 |
- |
1 |
334 |
|
Chr. 8 - 69749319 - 69749962 [-] |
GenBankmRNA |
BB750184.1 |
- |
1 |
133 |
|
Chr. 8 - 69749325 - 69749761 [-] |
View Full Product Details