Gene Transcripts
- Gene Symbol
- 4930429C20Rik
- Entrez Gene ID
- 73857
- Gene Name
- RIKEN cDNA 4930429C20 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.429392
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015232.1 |
- |
2 |
495 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY714786.1 |
- |
2 |
495 |
|
Chr. 14 - 71421012 - 71421613 [+] |
View Full Product Details