Gene Transcripts
- Gene Symbol
- Gm6634
- Entrez Gene ID
- 625901
- Gene Name
- predicted gene 6634
- Gene Aliases
- EG625901
- Chromosome Location
- Chr. 3: 70772379 - 70807291 on Build GRCm38
- UniGene ID
- Mm.366028
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040556.1 |
|
4 |
1265 |
|
GenBank mRNA |
AK144724.1 |
- |
4 |
1266 |
|
View Full Product Details