Gene Transcripts
- Gene Symbol
- 1700093C20Rik
- Entrez Gene ID
- 76623
- Gene Name
- RIKEN cDNA 1700093C20 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.460423
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007047.1 |
- |
1 |
728 |
|
View Full Product Details