Gene Transcripts
- Gene Symbol
- Kbtbd8os
- Entrez Gene ID
- 72217
- Gene Name
- kelch repeat and BTB (POZ) domain containing 8, opposite strand
- Gene Aliases
- 1700010K10Rik
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.396160
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005835.1 |
- |
2 |
402 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY705983.1 |
- |
2 |
400 |
|
Chr. 6 - 95114017 - 95117631 [-] |
View Full Product Details