Gene Transcripts
- Gene Symbol
- Zfp37
- Entrez Gene ID
- 22696
- Gene Name
- zinc finger protein 37
- Gene Aliases
- Tzn, Zfp-37
- Chromosome Location
- Chr. 4: 62189537 - 62208729 on Build GRCm38
- UniGene ID
- Mm.5011
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK043700.1 |
- |
1 |
304 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK081660.1 |
- |
|
1742 |
|
Chr. - - [-] |
View Full Product Details