Gene Transcripts
- Gene Symbol
- Agbl3
- Entrez Gene ID
- 76223
- Gene Name
- ATP/GTP binding protein-like 3
- Gene Aliases
- 2900053G10Rik, 4930431N21Rik, 6530406M24Rik, CCP3
- Chromosome Location
- Chr. 6: 34780432 - 34862184 on Build GRCm38
- UniGene ID
- Mm.150047
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015278.1 |
- |
1 |
371 |
|
View Full Product Details