Gene Transcripts
- Gene Symbol
- Gm10010
- Entrez Gene ID
- 434089
- Gene Name
- predicted gene 10010
- Gene Aliases
- ENSMUSG00000056771
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.101361
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK039686.1 |
- |
2 |
727 |
|
- Gene Symbol
- Gm31138
- Entrez Gene ID
- 102633275
- Gene Name
- predicted gene, 31138
- Gene Aliases
- Chromosome Location
- Chr. 9: 3199856 - 3215379 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK133027.1 |
- |
5 |
1294 |
|
View Full Product Details